missing translation for 'onlineSavingsMsg'
Learn More
Learn More
trans-2-Pentenal 95.0+%, TCI America™
Supplier: TCI America P179025G
Specifications
| trans-2-Pentenal | |
| Yellow | |
| C5H8O | |
| 25 g | |
| trans-2-pentenal, 2-pentenal, e-2-pentenal, e-pent-2-enal, 3-ethylacrolein, pent-2-enal, 2-pentenal, e, 2-e-pentenal, 3-ethyl-2-propenal, e-pent-2-en-1-al | |
| CCC=CC=O | |
| 84.118 | |
| 84.12 | |
| Liquid |
| 1576-87-0 | |
| 81°C | |
| MFCD00009615 | |
| 1989 | |
| DTCCTIQRPGSLPT-ONEGZZNKSA-N | |
| (E)-pent-2-enal | |
| 5364752 | |
| ≥95.0% (GC) |
Chemical Identifiers
| 1576-87-0 | |
| 84.118 | |
| DTCCTIQRPGSLPT-ONEGZZNKSA-N | |
| 5364752 | |
| CCC=CC=O |
| C5H8O | |
| MFCD00009615 | |
| trans-2-pentenal, 2-pentenal, e-2-pentenal, e-pent-2-enal, 3-ethylacrolein, pent-2-enal, 2-pentenal, e, 2-e-pentenal, 3-ethyl-2-propenal, e-pent-2-en-1-al | |
| (E)-pent-2-enal |
Safety and Handling
RTECSNumber : SB1560000
TSCA : No
Recommended Storage : Refrigerator