missing translation for 'onlineSavingsMsg'
Learn More

trans-2-Pentenal 95.0+%, TCI America™

Catalog No. p-7135255
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P17905G 5 g
P179025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P17905G Supplier TCI America Supplier No. P17905G
Only null left

Chemical Identifiers

CAS 1576-87-0
Molecular Formula C5H8O
Molecular Weight (g/mol) 84.118
MDL Number MFCD00009615
InChI Key DTCCTIQRPGSLPT-ONEGZZNKSA-N
Synonym trans-2-pentenal, 2-pentenal, e-2-pentenal, e-pent-2-enal, 3-ethylacrolein, pent-2-enal, 2-pentenal, e, 2-e-pentenal, 3-ethyl-2-propenal, e-pent-2-en-1-al
PubChem CID 5364752
IUPAC Name (E)-pent-2-enal
SMILES CCC=CC=O

Specifications

Color Yellow
Boiling Point 81°C
Quantity 5 g
UN Number 1989
Formula Weight 84.12
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material trans-2-Pentenal
RTECSNumber SB1560000
TSCA No
Recommended Storage Refrigerator