missing translation for 'onlineSavingsMsg'
Learn More
Learn More
trans-2-Hexenoic Acid 98.0+%, TCI America™
$121.51 - $674.02
Chemical Identifiers
| CAS | 13419-69-7 |
|---|---|
| Molecular Formula | C6H9O2 |
| Molecular Weight (g/mol) | 113.14 |
| MDL Number | MFCD00002705 |
| InChI Key | NIONDZDPPYHYKY-SNAWJCMRSA-M |
| Synonym | trans-2-hexenoic acid, 2-hexenoic acid, e-hex-2-enoic acid, hex-2-enoic acid, trans-hex-2-enoic acid, e-2-hexenoic acid, hexenoic acid, 2e-hex-2-enoic acid, 2-hexenoic acid, 2e, 2-hexenoic acid, e |
| PubChem CID | 5282707 |
| ChEBI | CHEBI:87721 |
| IUPAC Name | (2E)-hex-2-enoate |
| SMILES | CCC\C=C\C([O-])=O |
Chemical Identifiers
| 13419-69-7 | |
| 113.14 | |
| NIONDZDPPYHYKY-SNAWJCMRSA-M | |
| 5282707 | |
| (2E)-hex-2-enoate |
| C6H9O2 | |
| MFCD00002705 | |
| trans-2-hexenoic acid, 2-hexenoic acid, e-hex-2-enoic acid, hex-2-enoic acid, trans-hex-2-enoic acid, e-2-hexenoic acid, hexenoic acid, 2e-hex-2-enoic acid, 2-hexenoic acid, 2e, 2-hexenoic acid, e | |
| CHEBI:87721 | |
| CCC\C=C\C([O-])=O |
Specifications
| 13419-69-7 | |
| 118°C | |
| MFCD00002705 | |
| trans-2-hexenoic acid, 2-hexenoic acid, e-hex-2-enoic acid, hex-2-enoic acid, trans-hex-2-enoic acid, e-2-hexenoic acid, hexenoic acid, 2e-hex-2-enoic acid, 2-hexenoic acid, 2e, 2-hexenoic acid, e | |
| CCC\C=C\C([O-])=O | |
| 113.14 | |
| CHEBI:87721 | |
| ≥98.0% (GC) | |
| trans-2-Hexenoic Acid |
| White-Yellow | |
| C6H9O2 | |
| 3261 | |
| NIONDZDPPYHYKY-SNAWJCMRSA-M | |
| (2E)-hex-2-enoate | |
| 5282707 | |
| 114.14 | |
| Crystals |
Safety and Handling
EINECSNumber : (2)-0609
TSCA : Yes