missing translation for 'onlineSavingsMsg'
Learn More
Learn More
trans-2-Hexenoic Acid 98.0+%, TCI America™
Supplier: TCI America H0383250G
| Quantity | 250 g |
|---|
Chemical Identifiers
| 13419-69-7 | |
| 113.14 | |
| NIONDZDPPYHYKY-SNAWJCMRSA-M | |
| 5282707 | |
| (2E)-hex-2-enoate |
| C6H9O2 | |
| MFCD00002705 | |
| trans-2-hexenoic acid, 2-hexenoic acid, e-hex-2-enoic acid, hex-2-enoic acid, trans-hex-2-enoic acid, e-2-hexenoic acid, hexenoic acid, 2e-hex-2-enoic acid, 2-hexenoic acid, 2e, 2-hexenoic acid, e | |
| CHEBI:87721 | |
| CCC\C=C\C([O-])=O |
Specifications
| trans-2-Hexenoic Acid | |
| White-Yellow | |
| C6H9O2 | |
| MFCD00002705 | |
| trans-2-hexenoic acid, 2-hexenoic acid, e-hex-2-enoic acid, hex-2-enoic acid, trans-hex-2-enoic acid, e-2-hexenoic acid, hexenoic acid, 2e-hex-2-enoic acid, 2-hexenoic acid, 2e, 2-hexenoic acid, e | |
| CCC\C=C\C([O-])=O | |
| 113.14 | |
| CHEBI:87721 | |
| ≥98.0% (GC) |
| 13419-69-7 | |
| 118°C | |
| 250 g | |
| 3261 | |
| NIONDZDPPYHYKY-SNAWJCMRSA-M | |
| (2E)-hex-2-enoate | |
| 5282707 | |
| 114.14 | |
| Crystals |
Safety and Handling
EINECSNumber : (2)-0609
TSCA : Yes