missing translation for 'onlineSavingsMsg'
Learn More

trans-2-Hexenoic Acid 98.0+%, TCI America™

Catalog No. H038325G
missing translation for 'changeView'
missing translation for 'orderingAttributeHoverText'
Quantity:
25 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H038325G 25 g
H0383250G 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 missing translation for 'options'
Catalog No. H038325G missing translation for 'mfr' TCI America missing translation for 'supplierNo' H038325G

Chemical Identifiers

CAS 13419-69-7
Molecular Formula C6H9O2
Molecular Weight (g/mol) 113.14
MDL Number MFCD00002705
InChI Key NIONDZDPPYHYKY-SNAWJCMRSA-M
Synonym trans-2-hexenoic acid, 2-hexenoic acid, e-hex-2-enoic acid, hex-2-enoic acid, trans-hex-2-enoic acid, e-2-hexenoic acid, hexenoic acid, 2e-hex-2-enoic acid, 2-hexenoic acid, 2e, 2-hexenoic acid, e
PubChem CID 5282707
ChEBI CHEBI:87721
IUPAC Name (2E)-hex-2-enoate
SMILES CCC\C=C\C([O-])=O

Specifications

Color White-Yellow
Boiling Point 118°C
Quantity 25 g
UN Number 3261
Formula Weight 114.14
Percent Purity ≥98.0% (GC)
Physical Form Crystals
Chemical Name or Material trans-2-Hexenoic Acid
EINECSNumber (2)-0609
TSCA Yes