missing translation for 'onlineSavingsMsg'
Learn More
Learn More
trans-2-Hexenal 97.0+%, TCI America™
$48.13 - $388.89
Chemical Identifiers
| CAS | 6728-26-3 |
|---|---|
| Molecular Formula | C6H10O |
| Molecular Weight (g/mol) | 98.145 |
| MDL Number | MFCD00007008 |
| InChI Key | MBDOYVRWFFCFHM-SNAWJCMRSA-N |
| Synonym | trans-2-hexenal, 2-hexenal, leaf aldehyde, e-2-hexenal, e-hex-2-enal, 2-trans-hexenal, beta-propyl acrolein, hexylenic aldehyde, hex-2-en-1-al, 2-hexenal, e |
| PubChem CID | 5281168 |
| ChEBI | CHEBI:28913 |
| IUPAC Name | (E)-hex-2-enal |
| SMILES | CCCC=CC=O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
H034525ML
|
TCI America
H034525ML |
25 mL |
Each for $48.13
|
|
|||||
|
H0345100ML
|
TCI America
H0345100ML |
100 mL |
Each for $117.44
|
|
|||||
|
H0345500ML
|
TCI America
H0345500ML |
500 mL |
Each for $388.89
|
|
|||||
Chemical Identifiers
| 6728-26-3 | |
| 98.145 | |
| MBDOYVRWFFCFHM-SNAWJCMRSA-N | |
| 5281168 | |
| (E)-hex-2-enal |
| C6H10O | |
| MFCD00007008 | |
| trans-2-hexenal, 2-hexenal, leaf aldehyde, e-2-hexenal, e-hex-2-enal, 2-trans-hexenal, beta-propyl acrolein, hexylenic aldehyde, hex-2-en-1-al, 2-hexenal, e | |
| CHEBI:28913 | |
| CCCC=CC=O |
Specifications
| 6728-26-3 | |
| 146°C | |
| MFCD00007008 | |
| 1988 | |
| MBDOYVRWFFCFHM-SNAWJCMRSA-N | |
| (E)-hex-2-enal | |
| 5281168 | |
| 98.15 | |
| Liquid |
| Yellow | |
| C6H10O | |
| 25 mL | |
| trans-2-hexenal, 2-hexenal, leaf aldehyde, e-2-hexenal, e-hex-2-enal, 2-trans-hexenal, beta-propyl acrolein, hexylenic aldehyde, hex-2-en-1-al, 2-hexenal, e | |
| CCCC=CC=O | |
| 98.145 | |
| CHEBI:28913 | |
| ≥97.0% (GC) | |
| trans-2-Hexenal |
Safety and Handling
EINECSNumber : (2)-0520
RTECSNumber : MP5900000
TSCA : Yes
Recommended Storage : Refrigerator