missing translation for 'onlineSavingsMsg'
Learn More
Learn More
trans-2-Hexenal 97.0+%, TCI America™
Supplier: TCI America H0345500ML
Specifications
| trans-2-Hexenal | |
| Yellow | |
| C6H10O | |
| 500 mL | |
| trans-2-hexenal, 2-hexenal, leaf aldehyde, e-2-hexenal, e-hex-2-enal, 2-trans-hexenal, beta-propyl acrolein, hexylenic aldehyde, hex-2-en-1-al, 2-hexenal, e | |
| CCCC=CC=O | |
| 98.145 | |
| CHEBI:28913 | |
| ≥97.0% (GC) |
| 6728-26-3 | |
| 146°C | |
| MFCD00007008 | |
| 1988 | |
| MBDOYVRWFFCFHM-SNAWJCMRSA-N | |
| (E)-hex-2-enal | |
| 5281168 | |
| 98.15 | |
| Liquid |
Chemical Identifiers
| 6728-26-3 | |
| 98.145 | |
| MBDOYVRWFFCFHM-SNAWJCMRSA-N | |
| 5281168 | |
| (E)-hex-2-enal |
| C6H10O | |
| MFCD00007008 | |
| trans-2-hexenal, 2-hexenal, leaf aldehyde, e-2-hexenal, e-hex-2-enal, 2-trans-hexenal, beta-propyl acrolein, hexylenic aldehyde, hex-2-en-1-al, 2-hexenal, e | |
| CHEBI:28913 | |
| CCCC=CC=O |
Safety and Handling
EINECSNumber : (2)-0520
RTECSNumber : MP5900000
TSCA : Yes
Recommended Storage : Refrigerator