missing translation for 'onlineSavingsMsg'
Learn More

trans-2-[3-(4-tert-Butylphenyl)-2-methyl-2-propenylidene]malononitrile 98.0+%, TCI America™

Catalog No. p-7123062
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B3635100MG 100 mg
B36351G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B3635100MG Supplier TCI America Supplier No. B3635100MG

Chemical Identifiers

CAS 300364-84-5
Molecular Formula C17H18N2
Molecular Weight (g/mol) 250.345
MDL Number MFCD03453260
InChI Key OIASAVWSBWJWBR-UKTHLTGXSA-N
Synonym DCTB
PubChem CID 6382826
ChEBI CHEBI:64343
IUPAC Name 2-[(E)-3-(4-tert-butylphenyl)-2-methylprop-2-enylidene]propanedinitrile
SMILES CC(=CC1=CC=C(C=C1)C(C)(C)C)C=C(C#N)C#N

Specifications

Melting Point 125°C
Color Yellow
Quantity 100 mg
UN Number 3439
Formula Weight 250.35
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material trans-2-[3-(4-tert-Butylphenyl)-2-methyl-2-propenylidene]malononitrile