CAS RN 300364-84-5
IUPAC Name:
2-[3-(4-tert-butylphenyl)-2-methylprop-2-en-1-ylidene]propanedinitrile
Synonyms:
2-[3-(4-tert-butylphenyl)-2-methylprop-2-en-1-ylidene]propanedinitrile
Molecular Weight (g/mol):
250.35
Molecular Formula:
C17H18N2
InChi Key:
OIASAVWSBWJWBR-UHFFFAOYSA-N
SMILES:
CC(C=C(C#N)C#N)=CC1=CC=C(C=C1)C(C)(C)C
1
–
1
of
1
results
1
trans-2-[3-(4-tert-Butylphenyl)-2-methyl-2-propenylidene]malononitrile 98.0+%, TCI America™
CAS: 300364-84-5 Molecular Formula: C17H18N2 Molecular Weight (g/mol): 250.345 MDL Number: MFCD03453260 InChI Key: OIASAVWSBWJWBR-UKTHLTGXSA-N Synonym: DCTB PubChem CID: 6382826 ChEBI: CHEBI:64343 IUPAC Name: 2-[(E)-3-(4-tert-butylphenyl)-2-methylprop-2-enylidene]propanedinitrile SMILES: CC(=CC1=CC=C(C=C1)C(C)(C)C)C=C(C#N)C#N