missing translation for 'onlineSavingsMsg'
Learn More

trans-1,4-Dibromo-2-butene, 97%

Catalog No. p-2692336
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC406460250 25 g Glass bottle
AC406461000 100 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC406460250 Supplier Thermo Scientific Chemicals Supplier No. 406460250

CAS: 821-06-7 | C4H6Br2 | 213.9 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 821-06-7
Molecular Formula C4H6Br2
Molecular Weight (g/mol) 213.9
MDL Number MFCD00000249
InChI Key RMXLHIUHKIVPAB-OWOJBTEDSA-N
Synonym 1,4-dibromo-2-butene, trans-1,4-dibromo-2-butene, e-1,4-dibromobut-2-ene, e-1,4-dibromo-2-butene, 1,4-dibromobut-2-ene, trans-1,4-dibromobut-2-ene, 2-butene, 1,4-dibromo, 2-butene, 1,4-dibromo-, 2e, 2e-1,4-dibromobut-2-ene, 1,4-dibromo-2-butylene
PubChem CID 641245
IUPAC Name (E)-1,4-dibromobut-2-ene
SMILES C(C=CCBr)Br

Specifications

Melting Point 48°C to 53°C
Color White to Beige
Boiling Point 205°C
Flash Point 113°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula BrCH2CHCHCH2Br
Quantity 25 g
Solubility Information Solubility in water: slightly soluble. Other solubilities: slightly soluble in chloroform,soluble in ethanol,acetone
Formula Weight 213.9
Percent Purity 97%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material trans-1, 4-Dibromo-2-butene, 97%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Causes serious eye damage.
Toxic if swallowed.
Causes severe skin burns and eye damage.
Toxic if inhaled.
Precautionary Statement GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 212-472-7
Recommended Storage Refrigerator +4°C

RUO – Research Use Only