missing translation for 'onlineSavingsMsg'
Learn More

trans-1,2-Bis(phenylsulfonyl)ethylene 98.0+%, TCI America™

Catalog No. p-7119129
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B15741G 1 g
B15745G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B15741G Supplier TCI America Supplier No. B15741G

Chemical Identifiers

CAS 963-16-6
Molecular Formula C14H12O4S2
Molecular Weight (g/mol) 308.366
MDL Number MFCD00066528
InChI Key YGBXMKGCEHIWMO-VAWYXSNFSA-N
PubChem CID 5367099
IUPAC Name [(E)-2-(benzenesulfonyl)ethenyl]sulfonylbenzene
SMILES C1=CC=C(C=C1)S(=O)(=O)C=CS(=O)(=O)C2=CC=CC=C2

Specifications

Melting Point 230°C
Color White-Yellow
Quantity 1 g
Formula Weight 308.37
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material trans-1,2-Bis(phenylsulfonyl)ethylene