accessibility menu, dialog, popup

UserName

OSOSOO(E)

CAS RN 963-16-6

IUPAC Name: [(1E)-2-(benzenesulfonyl)ethenesulfonyl]benzene
Synonyms: [(1E)-2-(benzenesulfonyl)ethenesulfonyl]benzene
Molecular Weight (g/mol): 308.37
Molecular Formula: C14H12O4S2
InChi Key: YGBXMKGCEHIWMO-VAWYXSNFSA-N
SMILES: O=S(=O)(\C=C\S(=O)(=O)C1=CC=CC=C1)C1=CC=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results