missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Tranilast, ACROS Organics™
$148.06 - $455.74
Specifications
| Chemical Name or Material | Tranilast |
|---|---|
| CAS | 53902-12-8 |
| Solubility | insoluble in water |
| Merck Index | 9646 |
| Melting Point | 166.2°C to 168.2°C |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AC468610010
|
Thermo Scientific™
468610010 |
1 g |
Each for $148.06
|
|
|||||
|
AC468610050
|
Thermo Scientific™
468610050 |
5 g |
Each for $455.74
|
|
|||||
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
| Tranilast | |
| insoluble in water | |
| 166.2°C to 168.2°C | |
| 327.33 | |
| White to Light-yellow to Green |
| 53902-12-8 | |
| 9646 | |
| C18 H17 N O5 | |
| Crystalline Powder |