missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Tranilast, ACROS Organics™

Catalog No. AC468610010
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC468610010 1 g
AC468610050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC468610010 Supplier Thermo Scientific™ Supplier No. 468610010

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Name or Material Tranilast
CAS 53902-12-8
Quantity 1 g
Solubility insoluble in water
Merck Index 9646
Melting Point 166.2°C to 168.2°C
Molecular Formula C18 H17 N O5
Formula Weight 327.33
Physical Form Crystalline Powder
Color White to Light-yellow to Green