missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TraceCERT™ Acetone- 2,4-DNPH, 1,000 μg/mL in Acetonitrile, MilliporeSigma™ Supelco™
$99.36 - $447.06
Specifications
| Packaging | Ampule |
|---|---|
| CAS | 1567-89-1 |
| Product Line | TraceCERT |
| Flash Point | 2.0°C - closed cup |
| Concentration | 1000 μg/mL in acetonitrile |
Description
Featured Industries: EnvironmentalSpecifications
| Ampule | |
| TraceCERT | |
| 1000 μg/mL in acetonitrile | |
| Acetone-2,4-dinitrophenylhydrazone solution | |
| 238.20 |
| 1567-89-1 | |
| 2.0°C - closed cup | |
| C9H10N4O4 | |
| Organic Pollutant Standard | |
| TraceCERT™ Analytical Standard |
TraceCERT is a registered trademark of Sigma-Aldrich Co. LLC