missing translation for 'onlineSavingsMsg'
Learn More

TraceCERT™ Acetone- 2,4-DNPH, 1,000 μg/mL in Acetonitrile, MilliporeSigma™ Supelco™

Catalog No. 111020349
Change view
Click to view available options
Quantity:
1 mL
5 x 1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111020349 1 mL
111018699 5 x 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 111020349 Supplier Merck Emd Millipore Supplier No. CRM47341

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Featured Industries: Environmental

Specifications

Chemical Name or Material Acetone-2,4-dinitrophenylhydrazone
Packaging Ampule
CAS 1567-89-1, 75-05-8
Product Line TraceCERT
Flash Point 2.0°C - closed cup
Concentration 1000 μg/mL in acetonitrile
Linear Formula C9H10N4O4
Technique Liquid Chromatography
Certifications/Compliance ISO/IEC 17025
Synonym Acetone-2,4-dinitrophenylhydrazone solution
Type Organic Pollutant Standard
Solvent or Matrix Acetonitrile
Industry Type Environmental Testing
Molecular Weight (g/mol) 238.20
Quantity 1 mL
Formula Weight 238.20
Grade TraceCERT™ Analytical Standard
Show More Show Less

TraceCERT is a registered trademark of Sigma-Aldrich Co. LLC