missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ Traceable™ Memory-Loc™ Ultra-Low Temperature USB Datalogging Refrigerator/Freezer Thermometer
Traceable™ Datalogging Thermometer that can be used in 21 CFR 11 environments
$858.61 - $880.48
Specifications
| Type | Temperature Datalogger |
|---|---|
| Display Type | LCD |
| Calibration | Calibrated |
| Alarm Types | LED Indicator, Audible, Flashing LCD Segment |
| Software Type | TraceableLIVE |
Description
Traceable™ Datalogging Thermometer that can be used in 21 CFR 11 environments
Fisher Scientific™ Traceable™ Memory-Loc™ Datalogging Thermometers can be used in 21 CFR 11 environments. High-accuracy thermometer has a temperature range of -90°C to 105°C (-130F to 221°F)and resolution of 0.01 and maintains an accuracy of ±0.25°C across the entire temperature range. Ideal for monitoring temperatures in refrigerators, freezers, water baths, heating blocks, and incubators.- Data-logging thermometer with fixed one-minute timing intervals
- Meets current CDC requirements and additional recommendations for vaccine storage and monitoring
- Can be used in 21 CFR 11 environments, raw data is locked and cannot be cleared or changed on base unit
- Stores up to ten unique alarm events
- Capable of storing over 1 million temperature observations
- Hassle-free retrieval of data; unit can remain in use while downloading and analyzing data
- Status indicators: Low battery, low memory, USB data transfer and active alarm state
- Traceable to NIST for accuracy
- High-impact, chemical-resistant ABS plastic case measures 7 x 10.8 x 1.9cm (2.75 x 4.25 x 0.75 in.)
- Weight: 5 oz.
- Alarm tracking feature stores in memory each alarm event detailing when the alarm state occurred, as well as when the unit returned back to within range
- Alarm state indicators include visual LEDs, audio alerts, and flashing LCD segments
- Data for up to ten different alarm events can be retrieved with the most current event viewable on the display
- All new feature allows recorded data (CSV file) to be transferred from thermometer to PC (or Mac™) using a USB flash drive (not included)
- No software is needed for computer interface
- Each thermometer has a unique ID allowing multiple units to be used in the same location
- Raw data on unit cannot be overwritten or modified
- Indicator light will appear when 95% of memory is full
- Can be used in environments requiring 21 CFR 11 compliance
- Thermometer logs two years of observations with a one-minute logging interval
- Device will stop recording when storage is full and will not overwrite recorded data
- Data is unable to be deleted from memory once stored
- Bottle probes are sealed in a miniature bottle (1 x 2.5 in.) filled with nontoxic glycol
- Solution is GRAS (generally recognized as safe) by the FDA (Food and Drug Administration)
- Eliminates concerns about incidental contact with food or drinking water
- Temperature-buffered bottle sensor eliminates rapid changes when refrigerator door is opened
- Ideal for use in monitoring storage conditions for vaccines or other important samples
- Meets all current CDC requirements and recommendations for temperature monitoring of vaccine storage as part of the Vaccines for Children program
- Temperature Range: -90°C to 105°C (-130F to 221°F)
- Temperature Resolution: 0.01°
- Accuracy: ±0.2°C
- Calibration Points: -80°C, -40°C, 0°C and 100°C
- Part Number 15-079-622 includes 1 316 Stainless Steel Probe with Platinum RTD Sensor
- Part Number 15-079-623 includes 2 316 Stainless Steel Probes with Platinum RTD Sensors
Specifications
| Temperature Datalogger | |
| Calibrated | |
| TraceableLIVE | |
| Platinum RTD, Temperature-Buffered Bottle Sensor | |
| -90°C to +105°C (-130°F to +221°F) | |
| -130°F to 221°F |
| LCD | |
| LED Indicator, Audible, Flashing LCD Segment | |
| USB | |
| ±0.2°C | |
| 0.01° | |
| -90°C to +105°C |