Learn More
Thermo Scientific™ TRACE™ TR-FAME GC Columns
For separation of cis/trans isomers and FAMEs with various degrees of unsaturation, choose this low-bleed, high-polarity column optimized for FAME analysis.
$1039.84 - $4509.60
Specifications
Product Line TRACE
Stationary Phase Trace™ TR-FAME
Max. Temperature 250/260°C
Polarity Polar
View More Specs
Products 9
Catalog Number Mfr. No. Length (Metric) Diameter (Metric) Price Quantity  
Catalog Number Mfr. No. Length (Metric) Diameter (Metric) Price Quantity  
26-0M0-96P
View Documents Promotion Details
Thermo Scientific™
260M096P
10 m 0.1 mm
Each for $1,039.84
Only null left
03-052-622
View Documents
Thermo Scientific™
260M238P
100 m 0.25 mm
Each for $3,769.12
Only null left
26-0M1-41P
View Documents Promotion Details
Thermo Scientific™
260M141P
30 m 0.22 mm
Each for $1,643.44
Only null left
03-150-239
View Documents
Thermo Scientific™
260M142P
30 m 0.25 mm
Each for $1,643.44
Only null left
26-0M1-43P
View Documents Promotion Details
Thermo Scientific™
260M143P
30 m 0.32 mm
Each for $1,832.94
Only null left
26-0M1-47P
View Documents
Thermo Scientific™
260M147P
50 m 0.22 mm
Each for $2,817.68
Only null left
03-150-244
View Documents
Thermo Scientific™
260M154P
60 m 0.25 mm
Each for $3,204.81
Only null left
26-0M1-55P
View Documents Promotion Details
Thermo Scientific™
260M155P
60 m 0.32 mm
Each for $2,956.48
Only null left
26-0M1-66L
View Documents
Thermo Scientific™
260M166L
120 m 0.25 mm
Each for $4,509.60
Only null left
Description
Description
Choose Thermo Scientific™ TRACE™ TR-FAME GC Columns for separation of cis/trans isomers and fatty acid methyl esters (FAMEs) with various degrees of unsaturation. The TRACE TR-FAME column is a cyanopropylphenyl-based phase specifically designed for the separation of FAMEs. It is also suitable for any application that requires a high-polarity column for use with MS detection.
Excellent Quality and Reproducibility
  • Low bleed for MS, even at elevated temperatures
  • Outstanding robustness for difficult separations
  • Reliable and reproducible performance
  • Polar phase, 70% cyanopropyl polysilphenylene-siloxane
  • Higher operating temperature than other columns
Applications:
  • FAMEs
  • FAMEs cis/trans isomers
Recommended Uses:
  • FAMEs
Similar To:
  • DB-23
  • BPX70
  • Rtx™-2330
  • SP™-2330
  • CP-Sil 88
  • SP™-2380
  • HP-23
  • VF-23ms
  • 007-23
  • AT™-Silar
  • PE-23
Specifications
Specifications
TRACE
Trace™ TR-FAME
250/260°C
Polar
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.