missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ UHPLC Filters

Catalog No. 03150302
Change view
Click to view available options
Type:
1.0 mm I.D. Replacement Filter Cartridge, 0.2 μm
2.1 mm I.D. Replacement Filter Cartridge, 0.2 μm
UHPLC Direct Connect Filter Holder
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type
03150302 1.0 mm I.D. Replacement Filter Cartridge, 0.2 μm
03150301 UHPLC Direct Connect Filter Holder
03150300 2.1 mm I.D. Replacement Filter Cartridge, 0.2 μm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 03150302 Supplier Thermo Scientific™ Supplier No. 22185

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Protects columns without compromising performance.

Sacrificing performance is not an option with Thermo Scientific™ UHPLC filters that protect columns for Hypersil GOLD™ 1.9μm and Syncronis 1.7μm columns. These UHPLC filters provide a low-volume filter cartridge design, maintain peak shape, and ensure minimal efficiency loss through dispersion.

Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter.

Specifications

Particle Size 0.2 μm
Type 1.0 mm I.D. Replacement Filter Cartridge, 0.2 μm
Diameter (Metric) Inner 1 mm
Product Line Thermo Scientific