missing translation for 'onlineSavingsMsg'
Learn More

Trace Metals 2 For Environmental Analysis, SPEX CertiPrep™

ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material

$866.38 - $866.38

Chemical Identifiers

CAS 7697-37-2
Molecular Formula HNO3
Molecular Weight (g/mol) 63.01
MDL Number MFCD00011349
InChI Key GRYLNZFGIOXLOG-UHFFFAOYSA-N
PubChem CID 944
ChEBI CHEBI:48107
IUPAC Name nitric acid
SMILES O[N+]([O-])=O
View More Specs

Includes: NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
WP-3-500
View Documents
SPEX CertiPrep Group LLC
WP3500
500 mL Triple-leached LDPE bottle
Each for $866.38
Only null left
 
Description

Description

Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation

AnalyteConcentration
Sb20μg/mL
Tl20μg/mL
Ag10μg/mL

Certifications

Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001

Compliance

GHS/OSHA Complaint

Specifications

Chemical Identifiers

7697-37-2
63.01
GRYLNZFGIOXLOG-UHFFFAOYSA-N
CHEBI:48107
O[N+]([O-])=O
HNO3
MFCD00011349
944
nitric acid

Specifications

Triple-leached LDPE bottle
Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis.
MFCD00011349
Certified Reference Material
O[N+]([O-])=O
nitric acid
63.01
944
Environmental
Trace Metals 2, Groundwater and Wastewater Pollution Control Check Standard
7697-37-2
HNO3
AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation
GRYLNZFGIOXLOG-UHFFFAOYSA-N
2% HNO3
Wastewater Testing
500 mL
CHEBI:48107
Components in 2% HNO3:
Sb    •20μg/mL
Ag    •10μg/mL
Tl    •20μg/mL
Videos
Safety and Handling

Safety and Handling

DOTInformation : Hazardous

Recommended Storage : Ambient

SDS
Documents

Documents

Product intended for laboratory use only