missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ TRACE™ GC Columns for Drugs of Abuse

Catalog No. 26AC497P
Change view
Click to view available options
Diameter (Metric):
0.2 mm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Diameter (Metric)
26AC497P 0.2 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 26AC497P Supplier Thermo Scientific™ Supplier No. 26AC497P

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Analyze common drugs of abuse with these GC columns and achieve high performance, sensitivity and reproducibility.

Analyze amphetamines, codeine, morphine, and THC with high performance and reproducibility using Thermo Scientific™ TRACE™ GC Columns for Drugs of Abuse.

Tailored Specifically for Drugs of Abuse:
  • TRACE TR-DoA 5MS: Widely used for the analysis and determination of a range of toxicological target compounds including amphetamines, codeine, and morphine
  • TRACE TR-DoA 35MS: Ideal for the determination of carboxy-THC because of its higher polarity, making it the recommended column for use in drug testing labs for the confirmation of THC
Excellent Quality and Reproducibility:
  • Low bleed even at elevated temperatures
  • Outstanding robustness for difficult separations
  • Reliable and reproducible performance

Specifications

For Use With (Application) GC/MS for Amphetamines, Codeine, and Morphine
Film Thickness 0.33 μm
Length (Metric) 15 m
Diameter (Metric) 0.2 mm
USP Type G42
Stationary Phase Trace™ TR-DoA 35 MS
Diameter (Metric) Inner 0.2 mm
Product Line TRACE
Max. Temperature (Metric) 360/370°C