missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ TOPO™ XL-2 Complete PCR Cloning Kit, with One Shot™ OmniMAX™ 2 T1R Chemically Competent E. coli Cells

Catalog No. p-6927085
Encompass
Change view
Click to view available options
No. of Reactions:
10 Reactions
20 Reactions
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. No. of Reactions
K805010 10 Reactions
K805020 20 Reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
For Research Use Only. Not for use in diagnostic procedures.
Catalog No. K805010 Supplier Invitrogen™ Supplier No. K805010
Only null left
For Research Use Only. Not for use in diagnostic procedures.

Includes

Box 1:

  • pCR XL-2-TOPO vector
  • Salt Solution
  • T3 Primer
  • T7 Primer
  • XL Control PCR Template
  • XL Control PCR Primers
  • Platinum SuperFi Green PCR Master Mix
  • SuperFi GC Enhancer
  • Nuclease-free water
Box 2:
  • Gel Solubilization Buffer (L3)
  • Binding Buffer (B2)
  • Wash Buffer (W1)
  • Elution Buffer (E1)
  • PureLink Clean-up Spin Columns (in Wash Tubes)
  • PureLink Recovery/Elution Tubes
Box 3:
  • One Shot OmniMAX 2 T1R chemically competent E. coli
  • S.O.C. Medium
  • pUC 19 Control DNA

Provides all the necessary elements for efficient cloning of extra-long PCR products (up to 13 kb)

The kit uses the linearized and topoisomerase 1-activated pCR™-XL-2-TOPO vector, which is compatible with the cloning of blunt-end PCR fragments.  Amplification of long PCR fragments is enabled by Platinum™SuperFi™ Green PCR Master Mix, which is included in the kit.  Topoisomerase I activation of the vector allows PCR products to be ligated in just 5 minutes on your bench–top, resulting in high cloning efficiency (up to 90% positive recombinants with 10 kb fragments).

The pCR-XL-2-TOPO vector includes:
  • ccdB gene for positive selection
  • EcoR I site flanking the PCR product insertion site for easy excision of inserts
  • Ampicillin- and kanamycin-resistance genes for your choice of selection in E. coli
  • T7 promoter/priming site for in vitro RNA transcription and sequencing
  • T7, T3, and M13 forward and reverse primer sites for sequencing
The TOPO XL-2 Complete PCR Cloning Kit includes:
  • TOPO XL-2 PCR Cloning Kits containing the pCR-XL-2-TOPO vector
  • Platinum SuperFi Green PCR Master Mix—featuring a proofreading DNA polymerase (>100X fidelity compared to Taq) with high processivity and a density gradient to generate accurate, long PCR amplicons ready to load onto an agarose gel for gel extraction
  • PureLink™ Quick Gel Extraction and PCR Purification Combo Kit—designed to purify DNA fragments from agarose gels or direct PCR purification using a silica-based spin cartridge in < 30 minutes
  • One Shot™ OmniMAX™ 2 T1R Chemically Competent E. coli Cells—an improved chemically competent cell line, perfect for use in all cloning applications.

Specifications

Bacterial or Yeast Strain OmniMAX2
Antibiotic Resistance Bacterial Ampicillin (AmpR), Kanamycin (KanR)
Cell Type Chemically Competent E. coli
Cloning Method Zero Blunt TOPO
Content And Storage Box 1: Store at -5 to -30°C.
• pCR XL-2-TOPO vector
• Salt Solution
• T3 Primer
• T7 Primer
• XL Control PCR Template
• XL Control PCR Primers
• Platinum SuperFi Green PCR Master Mix
• SuperFi GC Enhancer
• Nuclease-free water

Box 2: Store at room temperature.
• Gel Solubilization Buffer (L3)
• Binding Buffer (B2)
• Wash Buffer (W1)
• Elution Buffer (E1)
• PureLink Clean-up Spin Columns (in Wash Tubes)
• PureLink Recovery/Elution Tubes

Box 3:
• One Shot OmniMAX 2 T1R chemically competent E. coli
• S.O.C. Medium
• pUC 19 Control DNA

Store competent cells and pUC19 at -65 to -85°C.

Format Kit
For Use With (Application) PCR Cloning
No. of Reactions 10 Reactions
Product Line TOPO
Product Type Cloning Kit
Promoter T3, T7
Quantity 10 reactions
Shipping Condition Approved for shipment at Room Temperature or on Wet or Dry Ice
Vector pCR-XL-2-TOPO
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.