missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Toluidine Blue, 1% Aqueous, Certified, LabChem™
$156.75 - $900.14
Chemical Identifiers
| CAS | 92-31-9 |
|---|---|
| Molecular Formula | C15H16ClN3S |
| Molecular Weight (g/mol) | 305.82 |
| MDL Number | MFCD00011934 |
| InChI Key | HNONEKILPDHFOL-UHFFFAOYSA-M |
| PubChem CID | 7083 |
| IUPAC Name | 3-amino-7-(dimethylamino)-2-methyl-5λâ´-phenothiazin-5-ylium chloride |
| SMILES | [Cl-].CN(C)C1=CC=C2N=C3C=C(C)C(N)=CC3=[S+]C2=C1 |
Chemical Identifiers
| 92-31-9 | |
| 305.82 | |
| HNONEKILPDHFOL-UHFFFAOYSA-M | |
| 3-amino-7-(dimethylamino)-2-methyl-5λâ´-phenothiazin-5-ylium chloride |
| C15H16ClN3S | |
| MFCD00011934 | |
| 7083 | |
| [Cl-].CN(C)C1=CC=C2N=C3C=C(C)C(N)=CC3=[S+]C2=C1 |
Specifications
| Passes Test | |
| 99,1 | |
| C15H16ClN3S | |
| Soluble in water | |
| HNONEKILPDHFOL-UHFFFAOYSA-M | |
| 3-amino-7-(dimethylamino)-2-methyl-5λâ´-phenothiazin-5-ylium chloride | |
| 7083 | |
| Certified | |
| Nitrogen oxides; Carbon monoxide; Carbon dioxide; Sulfur compounds | |
| 500 mL | |
| Toluidine Blue, 1% Aqueous |
Safety and Handling
GHS H Statement
Solution is not hazardous.
GHS P Statement
Wear eye protection, protective gloves.
Wash exposed skin thoroughly after handling.
If on skin: Wash with plenty of soap and water.
Take off contaminated clothing.
If skin irritation occurs: Get medical advice/attention.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
If eye irritation persists: Get medical advice/attention.
Recommended Storage : Room Temperature