missing translation for 'onlineSavingsMsg'
Learn More

Toluidine Blue, 1% Aqueous, Certified, LabChem™

Supplier:  LabChem LC261652

 View more versions of this product

Catalog No. LC261652


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Passes Test
1% Aqueous
99,1
C15H16ClN3S
Soluble in water
HNONEKILPDHFOL-UHFFFAOYSA-M
3-amino-7-(dimethylamino)-2-methyl-5λ⁴-phenothiazin-5-ylium chloride
7083
Certified
Nitrogen oxides; Carbon monoxide; Carbon dioxide; Sulfur compounds
1 L
Toluidine Blue
92-31-9,7732-18-5
C15H16ClN3S
MFCD00011934
Passes Test
[Cl-].CN(C)C1=CC=C2N=C3C=C(C)C(N)=CC3=[S+]C2=C1
305.82
305.82
Poly Bottle
Purple
Liquid

Chemical Identifiers

92-31-9
305.82
HNONEKILPDHFOL-UHFFFAOYSA-M
3-amino-7-(dimethylamino)-2-methyl-5λ⁴-phenothiazin-5-ylium chloride
C15H16ClN3S
MFCD00011934
7083
[Cl-].CN(C)C1=CC=C2N=C3C=C(C)C(N)=CC3=[S+]C2=C1
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
Solution is not hazardous.

GHS P Statement
Wear eye protection, protective gloves.
Wash exposed skin thoroughly after handling.
If on skin: Wash with plenty of soap and water.
Take off contaminated clothing.
If skin irritation occurs: Get medical advice/attention.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
If eye irritation persists: Get medical advice/attention.

Recommended Storage : Room Temperature

SDS
Documents

Documents

Promotions

Promotions