missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Toluene-3,4-dithiol 97.0+%, TCI America™
Supplier: TCI America T026625G
Specifications
| Toluene-3,4-dithiol | |
| White-Yellow | |
| C7H8S2 | |
| 25 g | |
| NIAAGQAEVGMHPM-UHFFFAOYSA-N | |
| 4-methylbenzene-1,2-dithiol | |
| 10334 | |
| ≥97.0% (GC,T) |
| 496-74-2 | |
| 137°C | |
| MFCD00004844 | |
| toluene-3,4-dithiol, dithiol, 1,2-benzenedithiol, 4-methyl, 3,4-dimercaptotoluene, 3,4-dimercaptotoluol, 1,2-dimercapto-4-methylbenzene, 4-methyl-1,2-benzenedithiol, 3,4-toluenedithiol, o-toluenesulfonylamide, usaf b-59 | |
| CC1=CC(=C(C=C1)S)S | |
| 156.261 | |
| 156.26 | |
| Crystalline Lumps |
Chemical Identifiers
| 496-74-2 | |
| 156.261 | |
| NIAAGQAEVGMHPM-UHFFFAOYSA-N | |
| 10334 | |
| CC1=CC(=C(C=C1)S)S |
| C7H8S2 | |
| MFCD00004844 | |
| toluene-3,4-dithiol, dithiol, 1,2-benzenedithiol, 4-methyl, 3,4-dimercaptotoluene, 3,4-dimercaptotoluol, 1,2-dimercapto-4-methylbenzene, 4-methyl-1,2-benzenedithiol, 3,4-toluenedithiol, o-toluenesulfonylamide, usaf b-59 | |
| 4-methylbenzene-1,2-dithiol |
Safety and Handling
RTECSNumber : XT2275000
TSCA : Yes