accessibility menu, dialog, popup

UserName

CH3SHHS

CAS RN 496-74-2

IUPAC Name: 4-methylbenzene-1,2-dithiol
Synonyms: 1,2-benzenedithiol, 4-methyl-
Molecular Weight (g/mol): 156.26
Molecular Formula: C7H8S2
InChi Key: NIAAGQAEVGMHPM-UHFFFAOYSA-N
SMILES: CC1=CC=C(S)C(S)=C1
Boiling Point 
  • (3)
  • (2)
Molecular Weight (g/mol) 
  • (3)
  • (2)
Percent Purity 
  • (3)
  • (2)
Physical Form 
  • (2)
  • (3)
Quantity 
  • (2)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (2)
  • (3)
  • (2)
  • (3)
  • (2)
  • (3)
  • (2)
  • (2)
  • (3)
  • (2)
  • (1)
  • (2)