missing translation for 'onlineSavingsMsg'
Learn More

Toluene-3,4-dithiol 97.0+%, TCI America™

Catalog No. T02661G
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T02661G 1 g
T02665G 5 g
T026625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. T02661G Supplier TCI America Supplier No. T02661G

Chemical Identifiers

CAS 496-74-2
Molecular Formula C7H8S2
Molecular Weight (g/mol) 156.261
MDL Number MFCD00004844
InChI Key NIAAGQAEVGMHPM-UHFFFAOYSA-N
Synonym toluene-3,4-dithiol, dithiol, 1,2-benzenedithiol, 4-methyl, 3,4-dimercaptotoluene, 3,4-dimercaptotoluol, 1,2-dimercapto-4-methylbenzene, 4-methyl-1,2-benzenedithiol, 3,4-toluenedithiol, o-toluenesulfonylamide, usaf b-59
PubChem CID 10334
IUPAC Name 4-methylbenzene-1,2-dithiol
SMILES CC1=CC(=C(C=C1)S)S

Specifications

Color White-Yellow
Boiling Point 137°C
Quantity 1 g
Formula Weight 156.26
Percent Purity ≥97.0% (GC,T)
Physical Form Crystalline Lumps
Chemical Name or Material Toluene-3,4-dithiol
RTECSNumber XT2275000
TSCA Yes