missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Specifications
| Tofisopam | |
| 157°C | |
| C22H26N2O4 | |
| 250 mg | |
| 1-(3,4-Dimethoxyphenyl)-5-ethyl-7,8-dimethoxy-4-methyl-5H-benzo[d][1,2]diazepine, 1-(3,4-Dimethoxyphenyl)-5-ethyl-7,8-dimethoxy-4-methyl-5H-2,3-benzodiazepine | |
| CCC1C(=NN=C(C2=CC(=C(C=C12)OC)OC)C3=CC(=C(C=C3)OC)OC)C | |
| 382.46 | |
| 382.46 | |
| Crystalline Powder |
| 22345-47-7 | |
| White-Yellow | |
| MFCD00823171 | |
| 3077 | |
| RUJBDQSFYCKFAA-UHFFFAOYSA-N | |
| 1-(3,4-dimethoxyphenyl)-5-ethyl-7,8-dimethoxy-4-methyl-5H-2,3-benzodiazepine | |
| 5502 | |
| ≥98.0% (HPLC) |
Chemical Identifiers
| 22345-47-7 | |
| 382.46 | |
| RUJBDQSFYCKFAA-UHFFFAOYSA-N | |
| 5502 | |
| CCC1C(=NN=C(C2=CC(=C(C=C12)OC)OC)C3=CC(=C(C=C3)OC)OC)C |
| C22H26N2O4 | |
| MFCD00823171 | |
| 1-(3,4-Dimethoxyphenyl)-5-ethyl-7,8-dimethoxy-4-methyl-5H-benzo[d][1,2]diazepine, 1-(3,4-Dimethoxyphenyl)-5-ethyl-7,8-dimethoxy-4-methyl-5H-2,3-benzodiazepine | |
| 1-(3,4-dimethoxyphenyl)-5-ethyl-7,8-dimethoxy-4-methyl-5H-2,3-benzodiazepine |
Safety and Handling
TSCA : No