accessibility menu, dialog, popup

UserName

H3CNNOH3COH3COH3COH3CCH3

CAS RN 22345-47-7

IUPAC Name: 1-(3,4-dimethoxyphenyl)-5-ethyl-7,8-dimethoxy-4-methyl-5H-2,3-benzodiazepine
Synonyms: tofisopam emandaxin tofizopam seriel tofisopamum grandaxin
Molecular Weight (g/mol): 382.46
Molecular Formula: C22H26N2O4
InChi Key: RUJBDQSFYCKFAA-UHFFFAOYNA-N
SMILES: CCC1C2=CC(OC)=C(OC)C=C2C(=NN=C1C)C1=CC=C(OC)C(OC)=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results