missing translation for 'onlineSavingsMsg'
Learn More

TOC Standard, 1000mg/L Carbon Stabilized with Hydrochloric Acid, Reagents

Catalog No. 01333422
Change view
Click to view available options
Quantity:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01333422 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01333422 Supplier Labchem Inc Supplier No. CT1433001C

TOC Standard, 1000mg/L Carbon Stabilized with Hydrochloric Acid, Reagent, use for Water and wastewater. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 7732-18-5 , 877-24-7
Molecular Formula H2O
Molecular Weight (g/mol) 18.02
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
IUPAC Name water
SMILES O

Specifications

Packaging Glass Bottle
Specific Gravity 1.001
Physical Form Liquid
Color Colorless
Boiling Point 100°C
For Use With (Equipment) Water Analyzer
Solubility Completely miscible
Quantity 1 L
Grade Reagent
Chemical Name or Material TOC Standard, 1000mg/L Carbon Stabilized with Hydrochloric Acid