missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tobramycin Sulfate USP, Medisca™
$371.07 - $2021.04
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
18-048-056
|
Medisca Pharmaceutique Inc
031906 |
1 g |
Each for $371.07
|
|
|||||
|
18-048-109
|
Medisca Pharmaceutique Inc
031903* |
5 g |
Each for $1,199.13
|
|
|||||
|
18-048-135
|
Medisca Pharmaceutique Inc
031901 |
10 g |
Each for $2,021.04
|
|
|||||
Chemical Identifiers
| 79645-27-5 |
Specifications
| α-Carboxybenzyl Penicillin Disodium Salt | |
| 79645-27-5 | |
| 1 g | |
| USP |
| Crystalline Powder | |
| White to Light Yellow | |
| 634 to 739 μg of Tobramycin/mg |