missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium Ferrocyanide, 7% (w/v) Aqueous Solution, Spectrum™ Chemical
Spécifications
Spécifications
| Physical Form | Solution |
| Quantity | 4 L |
| Packaging | Amber Glass Bottle |
| Chemical Name or Material | Potassium Ferrocyanide |
CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.