missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals TMB QUICK, for ELISA, Liquid Substrate

Catalog No. AC328051000
Change view
Click to view available options
Substrate Quantity:
50mL
100mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Substrate Quantity
AC328051000 100mL
AC328050500 50mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC328051000 Supplier Thermo Scientific Chemicals Supplier No. 328051000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Boiling Point 100°C
Color Clear
Form Liquid
Quantity 100mL
Substrate Type TMB Substrate
Packaging Glass Bottle
Substrate Quantity 100mL

RUO – Research Use Only