missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals TMB QUICK, for ELISA, Liquid Substrate

Supplier:  Thermo Scientific Chemicals 328050500

Substrate Quantity 50mL

 View more versions of this product

Catalog No. AC328050500

Only null left

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

100°C
Liquid
TMB Substrate
50mL
Clear
50mL
Glass Bottle
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions