missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals TMB EASY, Liquid Substrate

Supplier:  Thermo Scientific Chemicals 328031000

 View more versions of this product

Catalog No. AC328031000

Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

100°C
Liquid
TMB Substrate
4°C
Clear
100mL
Glass bottle
100mL
Product Suggestions

Product Suggestions

Videos
SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only