missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ TL 13-112
Description
Selective anaplastic lymphoma kinase (ALK) degrader (DC50 values are 10 and 40 nM in H3122 and Karpas 299 cells, respectively). Comprises the cereblon E3 ligase ligand Pomalidomide (Cat. No. 6302) conjugated to an ALK inhibitor. Inhibits proliferation of ALK-positive cancer cell lines. Maximum degradation exhibited at 16 h.
Negative control TL 13-110 (Cat. No. 6746) also available.
TL 13-112 is a selective anaplastic lymphoma kinase (ALK) Degrader (PROTAC®) (DC50 values are 10 and 40 nM in H3122 and Karpas 299 cells, respectively). Comprises the cereblon E3 ligase ligand Pomalidomide (Cat. No. 6302) conjugated to an ALK inhibitor. Inhibits proliferation of ALK-positive cancer cell lines. Maximum degradation exhibited at 16 h.
Negative control TL 13-110 (Cat. No. 6746) also available.
PROTAC® is a registered trademark of Arvinas Operations, Inc., and is used under license.
Specifications
Specifications
| Chemical Name or Material | N-[2-[2-[2-[4-[4-[[5-Chloro-4-[[2-[(1-methylethyl)sulfonyl]phenyl]amino]-2-pyrimidinyl]amino]-2-methyl-5-(1-methylethoxy)phenyl]-1-piperidinyl]ethoxy]ethoxy]ethyl]-2-[[2-(2,6-dioxo-3-piperidinyl)-2,3-dihydro-1,3-dioxo-1H-isoindol-4-yl]amino]acetamide |
| CAS | 2229037-19-6 |
| Quantity | 5 mg |
| Product Line | Tocris |
| Molecular Formula | C49H60ClN9O10S |
| Formula Weight | 1002.57 |
| Purity | 98% |