missing translation for 'onlineSavingsMsg'
Learn More

Titanium nitride sputtering target, 50.8mm (2.0in) dia x 6.35mm (0.250in) thick, 99.5% (metals basis)

Catalog No. AA41111KS
Change view
Click to view available options
Quantity:
1 Ea.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA41111KS 1 Ea.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AA41111KS Supplier Thermo Scientific Chemicals Supplier No. 041111KS

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 25583-20-4 | NTi | 61.87 g/mol

Titanium nitride is used in coating on titanium alloys, steel, carbide, and aluminum components to improve the substrate's surface properties. Used to harden and protect cutting and sliding surfaces, for decorative purposes. It is used to coat costume jewelry and automotive trim. It is also used in also used in microelectronics. It is also used in top layer coating, usually with nickel to chromium plated substrates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 25583-20-4
Molecular Formula NTi
Molecular Weight (g/mol) 61.87
MDL Number MFCD00049596
InChI Key NRTOMJZYCJJWKI-UHFFFAOYSA-N
Synonym titanium nitride, titanium nitride tin, nitridotitanium, unii-6rw464feff, titannitrid, titaniumylidyneamine, titanium mononitride, titanium nitride, <3 mum, 6rw464feff, titanium nitride tin grade a
PubChem CID 93091
IUPAC Name azanylidynetitanium
SMILES N#[Ti]

Specifications

Color Gray
Quantity 1 Ea.
Form Hot Pressed Material
Solubility Information Soluble in aqua regain. Insoluble in water.
Formula Weight 61.91
Percent Purity 99.5%
Assay (metals basis)
Chemical Name or Material Titanium nitride sputtering target, 50.8mm (2.0 in.) dia. x 6.35mm (0.250 in.) thick
EINECSNumber 247-117-5
RTECSNumber XR2230000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only