accessibility menu, dialog, popup

UserName

CAS RN 25583-20-4

IUPAC Name: titanioamine
Synonyms: titanium nitride
Molecular Weight (g/mol): 61.87
Molecular Formula: NTi
InChi Key: NRTOMJZYCJJWKI-UHFFFAOYSA-N
SMILES: N#[Ti]

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results