missing translation for 'onlineSavingsMsg'
Learn More

Titanium(IV) Chloride (ca. 19% in Toluene, ca. 1.0mol/L), TCI America™

Catalog No. T3238100ML
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T3238100ML 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T3238100ML Supplier TCI America Supplier No. T3238100ML

Laboratory reagent containing approximately 19% titanium(IV) chloride in toluene.

Chemical Identifiers

CAS 7550-45-0 , 108-88-3
Molecular Formula Cl4Ti
Molecular Weight (g/mol) 189.667
MDL Number MFCD00011267
InChI Key XJDNKRIXUMDJCW-UHFFFAOYSA-J
Synonym titanium iv chloride, titanium 4+ tetrachloride, acmc-1bjgv, titanium 4+ ion tetrachloride, parent, titanium +4 cation tetrachloride, titanium iv chloride 100ml, superlist names titanium chloride, t-4, titanic chloride; titanio tetracloruro di, unii-8o3pje5t7q; titanium chloride ticl4
PubChem CID 160960
IUPAC Name titanium(4+);tetrachloride
SMILES [Cl-].[Cl-].[Cl-].[Cl-].[Ti+4]

Specifications

Physical Form Liquid
Quantity 100 mL
UN Number 3286
Formula Weight 189.67
Percent Purity 19.0 to 22.0%
Grade Reagent
Chemical Name or Material Titanium(IV) Chloride (ca. 19% in Toluene, ca. 1.0mol/L)
RTECSNumber XR1925000
TSCA Yes
Recommended Storage Refrigerator