CAS RN 7550-45-0
Titanium(IV) chloride, 0.09M solution in 20% HCl, AcroSeal™
CAS: 7550-45-0 | Cl4Ti | 189.71 g/mol
Titanium(IV) chloride, 1M solution in dichloromethane, AcroSeal™
CAS: 7550-45-0 | Cl4Ti | 189.71 g/mol
Titanium(IV) chloride, 99.99% (metals basis)
CAS: 7550-45-0 Molecular Formula: Cl4Ti Molecular Weight (g/mol): 189.667 MDL Number: MFCD00011267 InChI Key: XJDNKRIXUMDJCW-UHFFFAOYSA-J Synonym: titanium iv chloride,titanium 4+ tetrachloride,acmc-1bjgv,titanium 4+ ion tetrachloride,parent,titanium +4 cation tetrachloride,titanium iv chloride 100ml,superlist names titanium chloride, t-4,titanic chloride; titanio tetracloruro di,unii-8o3pje5t7q; titanium chloride ticl4 PubChem CID: 160960 IUPAC Name: titanium(4+);tetrachloride SMILES: [Cl-].[Cl-].[Cl-].[Cl-].[Ti+4]
Titanium(IV) chloride, 1M soln. in dichloromethane
CAS: 7550-45-0 Molecular Formula: Cl4Ti Molecular Weight (g/mol): 189.667 MDL Number: MFCD00011267 InChI Key: XJDNKRIXUMDJCW-UHFFFAOYSA-J Synonym: titanium iv chloride,titanium 4+ tetrachloride,acmc-1bjgv,titanium 4+ ion tetrachloride,parent,titanium +4 cation tetrachloride,titanium iv chloride 100ml,superlist names titanium chloride, t-4,titanic chloride; titanio tetracloruro di,unii-8o3pje5t7q; titanium chloride ticl4 PubChem CID: 160960 IUPAC Name: titanium(4+);tetrachloride SMILES: [Cl-].[Cl-].[Cl-].[Cl-].[Ti+4]
Titanium(IV) chloride, 1M solution in toluene, AcroSeal™
CAS: 7550-45-0 | Cl4Ti | 189.71 g/mol
Titanium(IV) chloride, 99.9%
CAS: 7550-45-0 MDL Number: MFCD00011267 InChI Key: XJDNKRIXUMDJCW-UHFFFAOYSA-J Synonym: titanium iv chloride,titanium 4+ tetrachloride,acmc-1bjgv,titanium 4+ ion tetrachloride,parent,titanium +4 cation tetrachloride,titanium iv chloride 100ml,superlist names titanium chloride, t-4,titanic chloride; titanio tetracloruro di,unii-8o3pje5t7q; titanium chloride ticl4 PubChem CID: 160960 IUPAC Name: titanium(4+);tetrachloride SMILES: [Cl-].[Cl-].[Cl-].[Cl-].[Ti+4]
Titanium(IV) chloride, 99.0% min, typically 99.6%
CAS: 7550-45-0 Molecular Formula: Cl4Ti Molecular Weight (g/mol): 189.667 MDL Number: MFCD00011267 InChI Key: XJDNKRIXUMDJCW-UHFFFAOYSA-J Synonym: titanium iv chloride,titanium 4+ tetrachloride,acmc-1bjgv,titanium 4+ ion tetrachloride,parent,titanium +4 cation tetrachloride,titanium iv chloride 100ml,superlist names titanium chloride, t-4,titanic chloride; titanio tetracloruro di,unii-8o3pje5t7q; titanium chloride ticl4 PubChem CID: 160960 IUPAC Name: titanium(4+);tetrachloride SMILES: [Cl-].[Cl-].[Cl-].[Cl-].[Ti+4]
Titanium(IV) Chloride (ca. 19% in Toluene, ca. 1.0mol/L), TCI America™
CAS: 7550-45-0 Molecular Formula: Cl4Ti Molecular Weight (g/mol): 189.667 MDL Number: MFCD00011267 InChI Key: XJDNKRIXUMDJCW-UHFFFAOYSA-J Synonym: titanium iv chloride,titanium 4+ tetrachloride,acmc-1bjgv,titanium 4+ ion tetrachloride,parent,titanium +4 cation tetrachloride,titanium iv chloride 100ml,superlist names titanium chloride, t-4,titanic chloride; titanio tetracloruro di,unii-8o3pje5t7q; titanium chloride ticl4 PubChem CID: 160960 IUPAC Name: titanium(4+);tetrachloride SMILES: [Cl-].[Cl-].[Cl-].[Cl-].[Ti+4]
Titanium(IV) Chloride (14% in Dichloromethane, ca. 1.0mol/L), TCI America™
CAS: 7550-45-0 Molecular Formula: Cl4Ti Molecular Weight (g/mol): 189.667 InChI Key: XJDNKRIXUMDJCW-UHFFFAOYSA-J Synonym: titanium iv chloride,titanium 4+ tetrachloride,acmc-1bjgv,titanium 4+ ion tetrachloride,parent,titanium +4 cation tetrachloride,titanium iv chloride 100ml,superlist names titanium chloride, t-4,titanic chloride; titanio tetracloruro di,unii-8o3pje5t7q; titanium chloride ticl4 PubChem CID: 160960 IUPAC Name: titanium(4+);tetrachloride SMILES: [Cl-].[Cl-].[Cl-].[Cl-].[Ti+4]