missing translation for 'onlineSavingsMsg'
Learn More

TISAB IV solution, For determination of fluoride, MilliporeSigma™ Supelco™

Total ionic strength adjustment buffers (TISAB), is generally used for the microdetermination of fluorides.

$261.03 - $981.82

Specifications

CAS 64-19-7
Grade Determination of Fluoride
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
111002586
View Documents
Merck Emd Millipore
34574500ML
500 mL
Each for $261.03
Only null left
 
111002585
View Documents
Merck Emd Millipore
3457425L
2.5 L
Each for $981.82
Only null left
 
Description

Description

Specifications

Specifications

64-19-7
Determination of Fluoride
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'