missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tiron, Powder, Spectrum™ Chemical
$197.25 - $26842.96
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
18614462
|
Spectrum Chemical Mfg Cor
TI11510GM |
10 g | Amber Glass Bottle |
Each for $197.25
|
|
||||
|
18614464
|
Spectrum Chemical Mfg Cor
TI11525GM |
25 g | Amber Glass Bottle |
Each for $234.39
|
|
||||
|
18614463
|
Spectrum Chemical Mfg Cor
TI115100GM |
100 g | Amber Glass Bottle |
Each for $734.18
|
|
||||
|
18614466
|
Spectrum Chemical Mfg Cor
TI115500GM |
500 g | Amber Glass Bottle |
Each for $2,293.50
|
|
||||
|
18614461
|
Spectrum Chemical Mfg Cor
TI1151KG |
1 kg | Amber Glass Bottle |
Each for $3,098.39
|
|
||||
|
18614465
|
Spectrum Chemical Mfg Cor
TI11525KGBL |
25 kg | Amber Glass Bottle |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Chemical Identifiers
| 149-45-1 |
Specifications
| 149-45-1 | |
| Reagent | |
| White to Off-white | |
| Tiron |
| 1 | |
| Amber Glass Bottle | |
| 10 g |
CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.