missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tiron, Powder, Spectrum™ Chemical
Specifications
Specifications
| CAS Max % | 1 |
| Packaging | Amber Glass Bottle |
| Color | White to Off-white |
| Quantity | 500 g |
| Grade | Reagent |
| Chemical Name or Material | Tiron |
CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.