missing translation for 'onlineSavingsMsg'
Learn More

Tiron, Powder, Spectrum™ Chemical

Catalog No. 18614462
Change view
Click to view available options
Quantity:
10 g
25 g
100 g
500 g
1 kg
25 kg
Packaging:
Amber Glass Bottle
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18614462 10 g Amber Glass Bottle
18614464 25 g Amber Glass Bottle
18614463 100 g Amber Glass Bottle
18614466 500 g Amber Glass Bottle
18614461 1 kg Amber Glass Bottle
18614465 25 kg Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 18614462 Supplier Spectrum Chemical Mfg Cor Supplier No. TI11510GM

Chemical Identifiers

CAS 149-45-1

Specifications

CAS Max % 1
Grade Reagent
Packaging Amber Glass Bottle
Color White to Off-white
Quantity 10 g
Chemical Name or Material Tiron

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.