missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ Tiotropium bromide

Catalog No. 590250
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
590250 50 mg
590210 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 590250 Supplier Tocris Bioscience™ Supplier No. 5902/50

Potent muscarinic receptor antagonist

Tiotropium bromide is a potent muscarinic receptor antagonist. Inhibits electrical field stimulation-induced contraction of guinea pig trachea in vitro (IC50 = 0.17 nM). Induces smooth muscle relaxation and bronchodilation.

Specifications

Chemical Name or Material (1α,2β,4β,5α,7β)-7-[(2-Hydroxy-2,2-di-2-thienylacetyl)oxy]-9,9-dimethyl-3-oxa-9-azoniatricyclo[3.3.1.02,4]nonane bromide
CAS 136310-93-5
Synonym Ba 679 BR
Quantity 50 mg
Target Non-selective mAChR Antagonists
Molecular Formula C19H22BrNO4S2
Formula Weight 472.42
Purity >98%