missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tilorone Dihydrochloride 98.0+%, TCI America™
$164.03 - $661.75
Chemical Identifiers
| CAS | 27591-69-1 |
|---|---|
| Molecular Formula | C25H36Cl2N2O3 |
| Molecular Weight (g/mol) | 483.47 |
| MDL Number | MFCD00134071 |
| InChI Key | BSVYJQAWONIOOU-UHFFFAOYSA-N |
| Synonym | 2,7-Bis[2-(diethylamino)ethoxy]-9-fluorenone Dihydrochloride |
| PubChem CID | 33958 |
| IUPAC Name | dihydrogen 2,7-bis[2-(diethylamino)ethoxy]-9H-fluoren-9-one dichloride |
| SMILES | [H+].[H+].[Cl-].[Cl-].CCN(CC)CCOC1=CC=C2C3=CC=C(OCCN(CC)CC)C=C3C(=O)C2=C1 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
T2836100MG
|
TCI America
T2836100MG |
100 mg |
Each for $164.03
|
|
|||||
|
T28361G
|
TCI America
T28361G |
1 g |
Each for $661.75
|
|
|||||
Chemical Identifiers
| 27591-69-1 | |
| 483.47 | |
| BSVYJQAWONIOOU-UHFFFAOYSA-N | |
| 33958 | |
| [H+].[H+].[Cl-].[Cl-].CCN(CC)CCOC1=CC=C2C3=CC=C(OCCN(CC)CC)C=C3C(=O)C2=C1 |
| C25H36Cl2N2O3 | |
| MFCD00134071 | |
| 2,7-Bis[2-(diethylamino)ethoxy]-9-fluorenone Dihydrochloride | |
| dihydrogen 2,7-bis[2-(diethylamino)ethoxy]-9H-fluoren-9-one dichloride |
Specifications
| 27591-69-1 | |
| C25H36Cl2N2O3 | |
| 100 mg | |
| BSVYJQAWONIOOU-UHFFFAOYSA-N | |
| dihydrogen 2,7-bis[2-(diethylamino)ethoxy]-9H-fluoren-9-one dichloride | |
| 33958 | |
| ≥98.0% (HPLC,T) | |
| Tilorone Dihydrochloride |
| 234°C | |
| MFCD00134071 | |
| 2,7-Bis[2-(diethylamino)ethoxy]-9-fluorenone Dihydrochloride | |
| [H+].[H+].[Cl-].[Cl-].CCN(CC)CCOC1=CC=C2C3=CC=C(OCCN(CC)CC)C=C3C(=O)C2=C1 | |
| 483.47 | |
| 483.47 | |
| Crystalline Powder |
Safety and Handling
RTECSNumber : LL9000000
TSCA : No