accessibility menu, dialog, popup

UserName

CAS RN 27591-69-1

HHClClCH3NCH3OONCH3CH3O

CAS RN 27591-69-1

IUPAC Name: dihydrogen 2,7-bis[2-(diethylamino)ethoxy]-9H-fluoren-9-one dichloride
Synonyms: tilorone
Molecular Weight (g/mol): 483.47
Molecular Formula: C25H36Cl2N2O3
InChi Key: BSVYJQAWONIOOU-UHFFFAOYSA-N
SMILES: [H+].[H+].[Cl-].[Cl-].CCN(CC)CCOC1=CC=C2C3=CC=C(OCCN(CC)CC)C=C3C(=O)C2=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (1)
  • (1)