missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Tiapride hydrochloride

Catalog No. ac467200010
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC467200010 1 g
AC467200050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC467200010 Supplier Thermo Scientific Chemicals Supplier No. 467200010
Only null left

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 51012-33-0
Molecular Formula C15H24N2O4S·HCl

Specifications

Quantity 1 g
Solubility Solubility aq. soln.: slightly soluble. Other solubilities: soluble in acetic acid, slightly soluble in methanol
Melting Point 196.0°C to 199.0°C
Formula Weight 364.89
InChI Key OTFDPNXIVHBTKW-UHFFFAOYSA-N
SMILES [H+].[Cl-].CCN(CC)CCNC(=O)C1=CC(=CC=C1OC)S(C)(=O)=O
IUPAC Name hydrogen N-[2-(diethylamino)ethyl]-5-methanesulfonyl-2-methoxybenzamide chloride
Molecular Weight (g/mol) 364.89
Chemical Name or Material Tiapride hydrochloride
EINECSNumber 256-908-4

RUO – Research Use Only