missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Tianeptine, sodium salt

Catalog No. AC467052500
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC467052500 250 mg
AC467050010 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC467052500 Supplier Thermo Scientific Chemicals Supplier No. 467052500
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 30123-17-2
Molecular Formula C21H24ClN2NaO4S

Specifications

Quantity 250 mg
Melting Point 180.0°C
Formula Weight 458.93
Chemical Name or Material Tianeptine, sodium salt
EINECSNumber 250-059-3

RUO – Research Use Only