missing translation for 'onlineSavingsMsg'
Learn More

Tiamulin fumarate, 95%

Catalog No. AAJ66020MC
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ66020MC 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAJ66020MC Supplier Thermo Scientific Chemicals Supplier No. J66020MC

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 55297-96-6 | C{2}{8}H{4}{7}NO{4}S.C{4}H{4}O{4}, C32H51NO8S | 609.82 g/mol

Targets Gram-positive bacteria as well as a few mycoplasma species

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Targets Gram-positive bacteria as well as a few mycoplasma species

Specifications

CAS 55297-96-6
Synonym Thiamutilin fumarate
Quantity 100 mg
Melting Point 147°C to 148°C
Boiling Point 563°C
Molecular Formula C{2}{8}H{4}{7}NO{4}S.C{4}H{4}O{4}
Purity 95%